ENA369382077 Enamine

2-N,2-N,3-N-trimethylpyridine-2,3-diamine

  • MDL number MFCD17291922
  • Cas Number : 1341333-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101574
Availability details for
Quantity
  • Availability
  • Add to Cart