ENA369382089 Enamine

1-(4-bromobut-1-en-1-yl)-3-methoxybenzene

  • Cas Number : 1226484-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100197
Availability details for
Quantity
  • Availability
  • Add to Cart