ENA369382091 Enamine

1,2-oxazole-3-carbohydrazide

  • MDL number MFCD06738847
  • Cas Number : 62438-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-142
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101722
Availability details for
Quantity
  • Availability
  • Add to Cart