ENA369382093 Enamine

[1-(2-phenylethyl)pyrrolidin-2-yl]methanamine

  • MDL number MFCD10003525
  • Cas Number : 60737-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101724

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101724
Availability details for
Quantity
  • Availability
  • Add to Cart