ENA369382096 Enamine

4,4,4-trifluorobutanenitrile

  • MDL number MFCD02183570
  • Cas Number : 690-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101728

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-101728
pictograms GHS02,GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart