ENA369382101 Enamine

4-(4-methoxyphenyl)-1,7lambda6-dithia-4-azaspiro[4.4]nonane-3,7,7-trione

  • MDL number MFCD08143362
  • Cas Number : 880450-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101734
Availability details for
Quantity
  • Availability
  • Add to Cart