ENA369382102 Enamine

4-(2-phenylethyl)-1,7lambda6-dithia-4-azaspiro[4.4]nonane-3,7,7-trione

  • MDL number MFCD08143599
  • Cas Number : 879642-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101735
Availability details for
Quantity
  • Availability
  • Add to Cart