ENA369382103 Enamine

3-hydroxy-1$l^{6}-thiane-1,1-dione

  • MDL number MFCD00764774
  • Cas Number : 94158-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101736
Availability details for
Quantity
  • Availability
  • Add to Cart