ENA369382104 Enamine

4-(7-chloro-1,3-benzothiazol-2-yl)aniline

  • MDL number MFCD11637006
  • Cas Number : 1096285-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101737

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101737
Availability details for
Quantity
  • Availability
  • Add to Cart