ENA369382106 Enamine

2-(4-methoxypiperidin-1-yl)ethan-1-amine

  • MDL number MFCD06408770
  • Cas Number : 911300-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101739
Availability details for
Quantity
  • Availability
  • Add to Cart