ENA369382107 Enamine

N-[(2-fluorophenyl)methyl]bicyclo[2.2.1]heptan-2-amine hydrochloride

  • MDL number MFCD22196417
  • Cas Number : 1376042-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101740
Availability details for
Quantity
  • Availability
  • Add to Cart