ENA369382115 Enamine

2-(1,1-dioxo-1lambda6-thiolan-3-yl)-1,3-dioxo-2,3-dihydro-1H-isoindole-5-carboxylic acid

  • MDL number MFCD13623734
  • Cas Number : 1097046-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101776
Availability details for
Quantity
  • Availability
  • Add to Cart