ENA369382121 Enamine

3-(3-phenylpropoxy)pyrrolidine hydrochloride

  • MDL number MFCD09997067
  • Cas Number : 1185301-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101789

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101789
Availability details for
Quantity
  • Availability
  • Add to Cart