ENA369382123 Enamine

1-chloro-4-(3-chloroprop-1-yn-1-yl)benzene

  • MDL number MFCD12155851
  • Cas Number : 41412-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100203

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100203
Availability details for
Quantity
  • Availability
  • Add to Cart