ENA369382124 Enamine

1-(5-benzyl-1,2,4-oxadiazol-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD22196423
  • Cas Number : 1376344-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101793

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101793
Availability details for
Quantity
  • Availability
  • Add to Cart