ENA369382125 Enamine

2-(pyridin-3-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD11846890
  • Cas Number : 882032-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101794

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101794
Availability details for
Quantity
  • Availability
  • Add to Cart