ENA369382129 Enamine

5-(chlorosulfonyl)-2,3-difluorobenzoic acid

  • MDL number MFCD16817587
  • Cas Number : 847972-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101800

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101800
Availability details for
Quantity
  • Availability
  • Add to Cart