ENA369382133 Enamine

[1-(3-methylbutyl)piperidin-4-yl]methanamine

  • MDL number MFCD10003422
  • Cas Number : 1017452-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101807

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101807
Availability details for
Quantity
  • Availability
  • Add to Cart