ENA369382134 Enamine

5-(chloromethyl)-3-[(2-methylphenyl)methyl]-1,2,4-oxadiazole

  • MDL number MFCD16083311
  • Cas Number : 1247572-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101808

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101808
Availability details for
Quantity
  • Availability
  • Add to Cart