ENA369382141 Enamine

7-chloro-2,3,4,5-tetrahydro-1,5-benzothiazepine

  • MDL number MFCD20673094
  • Cas Number : 90346-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101844

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101844
Availability details for
Quantity
  • Availability
  • Add to Cart