ENA369382147 Enamine

2-azaspiro[4.6]undecane

  • MDL number MFCD06808629
  • Cas Number : 184-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101850
Availability details for
Quantity
  • Availability
  • Add to Cart