ENA369382153 Enamine

hexyl 4-aminobenzoate

  • MDL number MFCD00188946
  • Cas Number : 13476-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101858
Availability details for
Quantity
  • Availability
  • Add to Cart