ENA369382161 Enamine

2-chloro-5-(fluorosulfonyl)benzoic acid

  • MDL number MFCD00007418
  • Cas Number : 21346-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 339...341
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101873

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-101873
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart