ENA369382163 Enamine

1-(1-methyl-1H-pyrrol-3-yl)ethan-1-amine

  • MDL number MFCD07357480
  • Cas Number : 279226-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100019
Availability details for
Quantity
  • Availability
  • Add to Cart