ENA369382164 Enamine

propanebis(imidamide) dihydrochloride

  • MDL number MFCD00226873
  • Cas Number : 6623-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-101887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101887
Availability details for
Quantity
  • Availability
  • Add to Cart