ENA369382172 Enamine

1-cyclopropyl-2-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD22196436
  • Cas Number : 1376332-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101900
Availability details for
Quantity
  • Availability
  • Add to Cart