ENA369382180 Enamine

tert-butyl 2-propanoylpiperidine-1-carboxylate

  • MDL number MFCD20232809
  • Cas Number : 1334493-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101946

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101946
Availability details for
Quantity
  • Availability
  • Add to Cart