ENA369382190 Enamine

7-methyl-2,4,5,7-tetraazatricyclo[6.4.0.0,2,6]dodeca-1(8),3,5,9,11-pentaene-3-thiol

  • MDL number MFCD03767537
  • Cas Number : 2165-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-101962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101962
Availability details for
Quantity
  • Availability
  • Add to Cart