ENA369382194 Enamine

5-(4-fluorophenyl)pyrimidin-4-amine

  • MDL number MFCD07381981
  • Cas Number : 56239-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100239

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100239
Availability details for
Quantity
  • Availability
  • Add to Cart