ENA369382207 Enamine

3-(2-chloroacetyl)-1-(3-ethynylphenyl)urea

  • MDL number MFCD11185622
  • Cas Number : 1094370-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100240
Availability details for
Quantity
  • Availability
  • Add to Cart