ENA369382212 Enamine

1-(methylamino)-3-(morpholin-4-yl)propan-2-ol

  • MDL number MFCD03990574
  • Cas Number : 186343-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101990

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101990
Availability details for
Quantity
  • Availability
  • Add to Cart