ENA369382214 Enamine

2-(pyridin-4-yl)imidazo[1,2-a]pyridin-3-amine

  • MDL number MFCD14540235
  • Cas Number : 1215991-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101992

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-101992
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart