ENA369382217 Enamine

4-bromo-1-(2,2-difluoroethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD16686987
  • Cas Number : 1247470-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102000

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102000
Availability details for
Quantity
  • Availability
  • Add to Cart