ENA369382224 Enamine

2-imino-3,3,7,7-tetramethyl-2H,3H,5H,6H,7H-imidazo[1,2-c]imidazolidin-5-one

  • MDL number MFCD22196447
  • Cas Number : 108483-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236...238
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102039

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102039
Availability details for
Quantity
  • Availability
  • Add to Cart