ENA369382229 Enamine

6-cyclopropyl-1-(4,6-dimethylpyrimidin-2-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD16558763
  • Cas Number : 1223386-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102048

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102048
Availability details for
Quantity
  • Availability
  • Add to Cart