ENA369382231 Enamine

3-hydroxy-1-methylpyrrolidin-2-one

  • MDL number MFCD15529680
  • Cas Number : 132996-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102052

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102052
Availability details for
Quantity
  • Availability
  • Add to Cart