ENA369382234 Enamine

2-(ethanesulfinyl)cyclohexan-1-amine

  • MDL number MFCD20380845
  • Cas Number : 1342273-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102058

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102058
Availability details for
Quantity
  • Availability
  • Add to Cart