ENA369382235 Enamine

tert-butyl 3-(hydroxymethyl)-3-(trifluoromethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD11848050
  • Cas Number : 1283720-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102059
Availability details for
Quantity
  • Availability
  • Add to Cart