ENA369382237 Enamine

2-methyl-6-phenylpiperidine

  • MDL number MFCD08692850
  • Cas Number : 116435-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102062

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102062
Availability details for
Quantity
  • Availability
  • Add to Cart