ENA369382239 Enamine

1-ethyl-4-ethynyl-1H-pyrazole

  • MDL number MFCD17281919
  • Cas Number : 573982-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102064
Availability details for
Quantity
  • Availability
  • Add to Cart