ENA369382241 Enamine

2-(3-tert-butyl-1,2,4-oxadiazol-5-yl)-4-chloroaniline

  • MDL number MFCD11119615
  • Cas Number : 1040008-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-102067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102067
Availability details for
Quantity
  • Availability
  • Add to Cart