ENA369382246 Enamine

3-methyl-4-(3-methylpyrrolidin-1-yl)benzonitrile

  • MDL number MFCD14601333
  • Cas Number : 1247450-79-8
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102082
Availability details for
Quantity
  • Availability
  • Add to Cart