ENA369382254 Enamine

3-[(3,4-dimethoxyphenyl)amino]-2,3-dihydro-1lambda6-thiophene-1,1-dione

  • MDL number MFCD09057525
  • Cas Number : 914217-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102091
Availability details for
Quantity
  • Availability
  • Add to Cart