ENA369382256 Enamine

3-[(4-methylphenyl)amino]-2,3-dihydro-1lambda6-thiophene-1,1-dione

  • MDL number MFCD00159148
  • Cas Number : 39565-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102093
Availability details for
Quantity
  • Availability
  • Add to Cart