ENA369382258 Enamine

7,7-difluoro-3-azabicyclo[4.1.0]heptane hydrochloride

  • MDL number MFCD22056345
  • Cas Number : 1376248-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-102
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100245
Availability details for
Quantity
  • Availability
  • Add to Cart