ENA369382260 Enamine

1-(2,6-dichlorophenyl)propan-1-one

  • MDL number MFCD18394766
  • Cas Number : 1261792-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-102098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102098
Availability details for
Quantity
  • Availability
  • Add to Cart