ENA369382266 Enamine

decahydroquinoline-1-sulfonyl chloride

  • MDL number MFCD12095780
  • Cas Number : 1216155-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102122
Availability details for
Quantity
  • Availability
  • Add to Cart