ENA369382269 Enamine

1,3-dimethyl-5-(pyridin-3-yloxy)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD11103683
  • Cas Number : 122816-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91...93
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102126

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102126
Availability details for
Quantity
  • Availability
  • Add to Cart