ENA369382271 Enamine

5-chloro-3-(4-nitrophenyl)-1,2,4-oxadiazole

  • MDL number MFCD12815761
  • Cas Number : 1183366-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100248
Availability details for
Quantity
  • Availability
  • Add to Cart