ENA369382280 Enamine

methyl 2-[(5-bromopyridin-2-yl)(methyl)amino]acetate

  • MDL number MFCD16161353
  • Cas Number : 1249621-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-102148

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-102148
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart